missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™

Catalog No. F07645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F07645G Supplier TCI America Supplier No. F07645G
Only null left

Specifications

CAS 207981-46-2
Color Yellow
Boiling Point 191°C
Molecular Formula C8H3ClF4O
Quantity 5 g
MDL Number MFCD00061156
UN Number 3265
Synonym 2-fluoro-5-trifluoromethyl benzoyl chloride, alpha,alpha,alpha,6-tetrafluoro-m-toluoyl chloride, pubchem20102, acmc-209vlj, ksc496g7l, 2-fluoro-5-trifluoromethylbenzoylchloride, 2-fluoro-5-trifluromethylbenzoyl chloride, 5-trifluoromethyl-2-fluorobenzoyl chloride, 2-fluoro-5-trifluoromethyl benzoylchloride, 2-fluoro-5-trifluoromethyl-benzoyl chloride
InChI Key OFVKAIZITGCCDN-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)C(=O)Cl)F
IUPAC Name 2-fluoro-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol) 226.555
PubChem CID 2734880
Formula Weight 226.55
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzoyl Chloride
Show More Show Less