missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrotoluene 97.0+%, TCI America™

Catalog No. p-7129584
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F027610G 10 g
F027625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F027610G Supplier TCI America Supplier No. F027610G

Chemical Identifiers

CAS 455-88-9
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007284
InChI Key XUCYJGMIICONES-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
PubChem CID 68001
IUPAC Name 1-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=CC=C1F)[N+]([O-])=O

Specifications

Melting Point 39°C
Boiling Point 99°C
Quantity 10 g
Formula Weight 155.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Fluoro-5-nitrotoluene
TSCA Yes
Recommended Storage Refrigerator