missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrotoluene, 98+%

Catalog No. AAA1464918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1464918 50 g
AAA1464909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1464918 Supplier Thermo Scientific Chemicals Supplier No. A1464918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-88-9 | C7H6FNO2 | 155.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-88-9
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007284
InChI Key XUCYJGMIICONES-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
PubChem CID 68001
IUPAC Name 1-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=CC=C1F)[N+]([O-])=O

Specifications

Melting Point 37°C to 40°C
Boiling Point 98°C to 101°C (13 mmHg)
Flash Point 105°C (221°F)
Quantity 50 g
UN Number UN2811
Beilstein 1940341
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-5-nitrotoluene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 207-251-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only