missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-nitropyridine, 98%
Description
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Melting Point | 17°C to 21°C |
| Color | Colorless to Yellow |
| Density | 1.41 g/mL |
| Boiling Point | 86°C to 87°C (7 mmHg)(literature) |
| Refractive Index | 1.525 |
| Quantity | 5 g |
| Beilstein | 120351 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 142.09 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 207-260-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only