accessibility menu, dialog, popup

UserName

ONOFN

CAS RN 456-24-6

IUPAC Name: 2-fluoro-5-nitropyridine
Synonyms: 2-fluoro-5-nitropyridine
Molecular Weight (g/mol): 142.09
Molecular Formula: C5H3FN2O2
InChi Key: XOZAJNLUAODXSP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)N=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)