missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitropyridine, 98%

Catalog No. AAH6421006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6421006 5 g
AAH6421014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6421006 Supplier Thermo Scientific Chemicals Supplier No. H6421006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 456-24-6 | C5H3FN2O2 | 142.089 g/mol

It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 456-24-6
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.089
MDL Number MFCD03095059
InChI Key XOZAJNLUAODXSP-UHFFFAOYSA-N
Synonym 6-fluoro-3-nitropyridine, pyridine, 2-fluoro-5-nitro, 2-fluor-5-nitropyridin, 2-fluoro-5-nitro-pyridine, abbypharma ap-15-5253, pubchem1202, acmc-1arnr, 5-nitro-2-fluoropyridine, 2-fluoro-5-nitro pyridine
PubChem CID 95264
IUPAC Name 2-fluoro-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])F

Specifications

Melting Point 17°C to 21°C
Color Colorless to Yellow
Density 1.41 g/mL
Boiling Point 86°C to 87°C (7 mmHg)(literature)
Refractive Index 1.525
Quantity 5 g
Beilstein 120351
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-nitropyridine
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-260-6
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only