missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitropyridine 98.0+%, TCI America™

Catalog No. f098325g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F098325G 25 g
F09835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F098325G Supplier TCI America Supplier No. F098325G

Chemical Identifiers

CAS 456-24-6
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.089
MDL Number MFCD03095059
InChI Key XOZAJNLUAODXSP-UHFFFAOYSA-N
Synonym 6-fluoro-3-nitropyridine, pyridine, 2-fluoro-5-nitro, 2-fluor-5-nitropyridin, 2-fluoro-5-nitro-pyridine, abbypharma ap-15-5253, pubchem1202, acmc-1arnr, 5-nitro-2-fluoropyridine, 2-fluoro-5-nitro pyridine
PubChem CID 95264
IUPAC Name 2-fluoro-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])F

Specifications

Melting Point 18°C
Color Yellow
Boiling Point 87°C
Quantity 25 g
Formula Weight 142.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-nitropyridine