missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzonitrile, 98+%

Catalog No. AAA1241703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1241703 1 g
AAA1241706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1241703 Supplier Thermo Scientific Chemicals Supplier No. A1241703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17417-09-3 | C7H3FN2O2 | 166.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17417-09-3
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.11
MDL Number MFCD00042299
InChI Key YLACBMHBZVYOAP-UHFFFAOYSA-N
Synonym benzonitrile, 2-fluoro-5-nitro, 3-cyano-4-fluoronitrobenzene, 2-fluoro-5-nitro-benzonitrile, 2-fluoro-5-nitrobenzenecarbonitrile, 2-fluoro-5-nitrobenzenenitrile, 3-cyano-4-fluoro-1-nitrobenzene, pubchem4792, intermediates-zcf02297, acmc-209e7w, 5-nitro-2-fluorobenzonitrile
PubChem CID 519417
IUPAC Name 2-fluoro-5-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(F)C(=C1)C#N

Specifications

Melting Point 70°C to 72°C
Quantity 1 g
UN Number UN3439
Beilstein 2048720
Formula Weight 166.11
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-5-nitrobenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only