missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-nitrobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America F039125G
Specifications
| 2-Fluoro-5-nitrobenzoic Acid | |
| 139°C | |
| MFCD00134238 | |
| 2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid | |
| OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O | |
| 185.11 | |
| 185.11 | |
| Crystalline Powder |
| 7304-32-7 | |
| C7H4FNO4 | |
| 25 g | |
| ICXSHFWYCHJILC-UHFFFAOYSA-N | |
| 2-fluoro-5-nitrobenzoic acid | |
| 280997 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 7304-32-7 | |
| 185.11 | |
| ICXSHFWYCHJILC-UHFFFAOYSA-N | |
| 280997 | |
| OC(=O)C1=CC(=CC=C1F)[N+]([O-])=O |
| C7H4FNO4 | |
| MFCD00134238 | |
| 2-fluoro-5-nitrobenzoic acid, 2-fluoro-5-nitrobenzoicacid, 2-fluoro-5-nitro-benzoic acid, benzoic acid, 2-fluoro-5-nitro, 2-fluoro-5-nitro-benzoicacid, pubchem1307, acmc-209op6, ksc377e3h, 2-fluoro-5-nitrobenzotc acid, 2-fluoro-5-nitro benzoic acid | |
| 2-fluoro-5-nitrobenzoic acid |
Safety and Handling
TSCA : No