missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylphenol, 97%

Catalog No. p-7024990
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2027503 1 g
AAB2027506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2027503 Supplier Thermo Scientific Chemicals Supplier No. B2027503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63762-79-8 | C7H7FO | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63762-79-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00190101
InChI Key XEHPMVZYZDQLDN-UHFFFAOYSA-N
Synonym phenol, 2-fluoro-5-methyl, 2-fluoro-5-methyl-phenol, 4-fluoro-3-hydroxytoluene, 6-fluoro-m-cresol, 6-fluoo-m-cresol, pubchem1498, acmc-209vyr, 2-fluoro-5-methyl phenol, 2-fluoranyl-5-methyl-phenol, ksc494c5d
PubChem CID 182387
IUPAC Name 2-fluoro-5-methylphenol
SMILES CC1=CC(=C(C=C1)F)O

Specifications

Density 1.155
Boiling Point 172°C to 174°C
Flash Point 67°C (152°F)
Refractive Index 1.5105
Quantity 1 g
UN Number UN3265
Beilstein 2433017
Formula Weight 126.13
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-methylphenol
Hazard Category H227-H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only