missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylaniline 98.0+%, TCI America™

Catalog No. p-7129732
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05285G 5 g
F052825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05285G Supplier TCI America Supplier No. F05285G

Chemical Identifiers

CAS 452-84-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007654
InChI Key QZUXMXZNVAJNSE-UHFFFAOYSA-N
Synonym 3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
PubChem CID 262970
IUPAC Name 2-fluoro-5-methylaniline
SMILES CC1=CC=C(F)C(N)=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 125.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methylaniline