missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methylaniline, 98%

Catalog No. AAL1341106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1341106 5 g
AAL1341114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1341106 Supplier Thermo Scientific Chemicals Supplier No. L1341106

CAS: 452-84-6 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 452-84-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00007654
InChI Key QZUXMXZNVAJNSE-UHFFFAOYSA-N
Synonym 3-amino-4-fluorotoluene, 6-fluoro-m-toluidine, 2-fluoro-5-methylphenylamine, 2-fluoro-5-methylbenzenamine, benzenamine, 2-fluoro-5-methyl, 2-fluoro-5-methylbenzeneamine, 2-fluoro-5-methyl-aniline, pubchem1507, 2-fluoro-5-metylaniline, acmc-209k1v
PubChem CID 262970
IUPAC Name 2-fluoro-5-methylaniline
SMILES CC1=CC=C(F)C(N)=C1

Specifications

Density 1.109
Boiling Point 80°C to 86°C (11 mmHg)
Flash Point 81°C (177°F)
Odor Characteristic
Refractive Index 1.533
Quantity 5 g
UN Number UN2810
Beilstein 2715995
Formula Weight 125.15
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-methylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only