missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-iodopyridine 98.0+%, TCI America™

Catalog No. p-7129729
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05231G 1 g
F05235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05231G Supplier TCI America Supplier No. F05231G

Chemical Identifiers

CAS 171197-80-1
Molecular Formula C5H3FIN
Molecular Weight (g/mol) 222.99
MDL Number MFCD03095301
InChI Key GYZNHUNWABYAPO-UHFFFAOYSA-N
Synonym 2-fluoro-5-iodo pyridine, pyridine, 2-fluoro-5-iodo, 2-fluoro-5-iodo-pyridine, abbypharma ap-30-0927, pubchem6324, 5-iodo-2-fluoropyridine, acmc-1bao4, ksc493k0d, 2-fluoro-5-iodopyridine, 2-fluoro-5-iodopyridine 1g
PubChem CID 2783179
IUPAC Name 2-fluoro-5-iodopyridine
SMILES FC1=CC=C(I)C=N1

Specifications

Melting Point 35°C
Quantity 1 g
Formula Weight 222.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-iodopyridine