accessibility menu, dialog, popup

UserName

CAS RN 171197-80-1

IUPAC Name: 2-fluoro-5-iodopyridine
Synonyms: 2-fluoro-5-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: GYZNHUNWABYAPO-UHFFFAOYSA-N
SMILES: FC1=CC=C(I)C=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results