missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-iodobenzoic Acid 98.0+%, TCI America™

Catalog No. F07861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07861G 1 g
F07865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07861G Supplier TCI America Supplier No. F07861G

Chemical Identifiers

CAS 124700-41-0
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD03094517
InChI Key QNNJHBNTHVHALE-UHFFFAOYSA-N
Synonym benzoic acid, 2-fluoro-5-iodo, 2-fluoro-5-iodobenzoicacid, 2-fluoro-5-iodo-benzoic acid, pubchem1385, acmc-1c24w, ksc174o6d, rarechem al bo 1479, buttpark 122\01-06, 5-iodo-2-fluorobenzoic acid, 2-fluoro-5-iodobenzoic acid
PubChem CID 2778925
IUPAC Name 2-fluoro-5-iodobenzoic acid
SMILES C1=CC(=C(C=C1I)C(=O)O)F

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 1 g
Formula Weight 266.01
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-iodobenzoic Acid