missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%

Catalog No. AC445030050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445030050 5 g
AC445030010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445030050 Supplier Thermo Scientific Chemicals Supplier No. 445030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1112208-82-8 | C13H16BFO3 | 250.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1112208-82-8
Molecular Formula C13H16BFO3
Molecular Weight (g/mol) 250.08
MDL Number MFCD11867877
InChI Key WHYMLQVABFAGEK-UHFFFAOYSA-N
Synonym 2-fluoro-5-formylphenylboronic acid pinacol ester, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 2-fluoro-5-formylphenylboronic acid, pinacol ester, 2-fluoro-5-formylphenyl boronic acid pinacol ester, 2-fluoro-5-formylbenzeneboronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-4-fluorobenzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzaldehyde, benzaldehyde, 4-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 46738018
IUPAC Name 4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
SMILES CC1(C)OB(OC1(C)C)C1=C(F)C=CC(C=O)=C1

Specifications

Melting Point 32°C to 37°C
Boiling Point 105°C to 113°C (0.1 mmHg)
Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 250.08
Percent Purity 95%
Chemical Name or Material 2-Fluoro-5-formylphenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only