missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-formylbenzonitrile 98.0+%, TCI America™

Catalog No. C26925G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26925G 5 g
C26921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26925G Supplier TCI America Supplier No. C26925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 218301-22-5
Molecular Formula C8H4FNO
Molecular Weight (g/mol) 149.124
MDL Number MFCD01863558
InChI Key MOFRJTLODZILCR-UHFFFAOYSA-N
Synonym 3-cyano-4-fluorobenzaldehyde, 4-fluoro-3-cyanobenzaldehyde, 2-fluoro-5-formyl-benzonitrile, benzonitrile, 2-fluoro-5-formyl, 2-fluoro-5-formylbenzenecarbonitrile, 2-fluor-5-formylbenzonitrile, 10014 2-fluoro-5-formyl benzonitrile, pubchem2252, acmc-1cfa9, fluoro-5-formylbenzonitrile
PubChem CID 2769582
IUPAC Name 2-fluoro-5-formylbenzonitrile
SMILES C1=CC(=C(C=C1C=O)C#N)F

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 149.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-formylbenzonitrile