missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-formylbenzonitrile, 99%

Catalog No. AC397940010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397940010 1 g
AC397940050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC397940010 Supplier Thermo Scientific Chemicals Supplier No. 397940010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 218301-22-5 | C8H4FNO | 149.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 218301-22-5
Molecular Formula C8H4FNO
Molecular Weight (g/mol) 149.12
MDL Number MFCD01863558
InChI Key MOFRJTLODZILCR-UHFFFAOYSA-N
Synonym 3-cyano-4-fluorobenzaldehyde, 4-fluoro-3-cyanobenzaldehyde, 2-fluoro-5-formyl-benzonitrile, benzonitrile, 2-fluoro-5-formyl, 2-fluoro-5-formylbenzenecarbonitrile, 2-fluor-5-formylbenzonitrile, 10014 2-fluoro-5-formyl benzonitrile, pubchem2252, acmc-1cfa9, fluoro-5-formylbenzonitrile
PubChem CID 2769582
IUPAC Name 2-fluoro-5-formylbenzonitrile
SMILES C1=CC(=C(C=C1C=O)C#N)F

Specifications

Melting Point 80°C to 84°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula FC6H3(CN)CHO
Quantity 1 g
Formula Weight 149.12
Percent Purity 99%
Chemical Name or Material 2-Fluoro-5-formylbenzonitrile
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only