missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F08161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F08161G Supplier TCI America Supplier No. F08161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 503309-11-3
Melting Point 116°C
Color White-Yellow
Molecular Formula C7H5BF4O2
Quantity 1 g
MDL Number MFCD03094995
Synonym 2-fluoro-4-trifluoromethyl phenylboronic acid, 2-fluoro-4-trifluoromethylphenylboronic acid, 2-fluoro-4-trifluoromethyl benzeneboronic acid, 2-fluoro-4-trifluoromethyl phenyl boronic acid, boronic acid, 2-fluoro-4-trifluoromethyl phenyl, pubchem1859, acmc-209klg, ksc269g4l, 4-borono-3-fluorobenzotrifluoride
InChI Key SSFSVKVWAURAAM-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-4-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 16414145
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less