missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)nicotinic acid, 97%

Catalog No. p-7031515
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32618MD 250 mg
AAH3261803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32618MD Supplier Thermo Scientific Chemicals Supplier No. H32618MD
Only null left

CAS: 1040681-74-0 | C7H3F4NO2 | 209.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1040681-74-0
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD10687403
InChI Key ABXMDMDQUVWALB-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl pyridine-3-carboxylic acid, 2-fluoro-4-trifluoromethyl pyridine-3-carboxylicacid, 2-fluoro-4-trifluoromethyl nicotinic acid, acmc-20anv9
PubChem CID 28307766
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=CN=C(C(=C1C(F)(F)F)C(=O)O)F

Specifications

Melting Point 91°C to 93°C
Quantity 250 mg
Formula Weight 209.1
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)nicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only