missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzeneboronic acid, 97%

Catalog No. AAH2878806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2878806 5 g
AAH2878803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2878806 Supplier Thermo Scientific Chemicals Supplier No. H2878806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 503309-11-3 | C7H5BF4O2 | 207.919 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 503309-11-3
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.919
MDL Number MFCD03094995
InChI Key SSFSVKVWAURAAM-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl phenylboronic acid, 2-fluoro-4-trifluoromethylphenylboronic acid, 2-fluoro-4-trifluoromethyl benzeneboronic acid, 2-fluoro-4-trifluoromethyl phenyl boronic acid, boronic acid, 2-fluoro-4-trifluoromethyl phenyl, pubchem1859, acmc-209klg, ksc269g4l, 4-borono-3-fluorobenzotrifluoride
PubChem CID 16414145
IUPAC Name [2-fluoro-4-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(F)(F)F)F)(O)O

Specifications

Melting Point 113°C to 115°C
Quantity 5 g
Formula Weight 207.92
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only