missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(pentafluorothio)aniline, 97%

Catalog No. AAH34099MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34099MD 250 mg
AAH3409903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34099MD Supplier Thermo Scientific Chemicals Supplier No. H34099MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1240257-25-3 | C6H5F6NS | 237.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1240257-25-3
Molecular Formula C6H5F6NS
Molecular Weight (g/mol) 237.16
MDL Number MFCD16652511
InChI Key VNFMUDAEGKTTOA-UHFFFAOYSA-N
Synonym 2-fluoro-4-pentafluorosulfur aniline, 2-fluoro-4-pentafluorothio aniline, 2-fluoro-4-pentafluoro-??-sulfanyl aniline, 2-fluoro-4-pentafluoro-$l^ 6
PubChem CID 66523512
IUPAC Name 2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)aniline
SMILES NC1=C(F)C=C(C=C1)S(F)(F)(F)(F)F

Specifications

Quantity 250 mg
UN Number UN2810
Formula Weight 237.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(pentafluorothio)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only