missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-nitrophenol 97.0+%, TCI America™
$34.66 - $102.03
Chemical Identifiers
| CAS | 403-19-0 |
|---|---|
| Molecular Formula | C6H4FNO3 |
| Molecular Weight (g/mol) | 157.1 |
| MDL Number | MFCD00051970 |
| InChI Key | ORPHLVJBJOCHBR-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t |
| PubChem CID | 9825 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])F)O |
Chemical Identifiers
| 403-19-0 | |
| 157.1 | |
| ORPHLVJBJOCHBR-UHFFFAOYSA-N | |
| 9825 |
| C6H4FNO3 | |
| MFCD00051970 | |
| 2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)O |
Specifications
| 403-19-0 | |
| Yellow | |
| MFCD00051970 | |
| 2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)O | |
| 9825 | |
| ≥97.0% (GC) | |
| 2-Fluoro-4-nitrophenol |
| 120°C | |
| C6H4FNO3 | |
| 5 g | |
| ORPHLVJBJOCHBR-UHFFFAOYSA-N | |
| 157.1 | |
| 157.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : No