missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog No. p-7052442
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3349403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3349403 Supplier Thermo Scientific Chemicals Supplier No. H3349403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 660432-43-9 | Molecular Formula: C7H6FNO3 | Molecular Weight: 171.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 660432-43-9
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.13
MDL Number MFCD11110168
InChI Key XYGZVFTWIGGORD-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrophenyl methanol, 2-fluoro-4-nitrobenzyl alcohol, benzenemethanol, 2-fluoro-4-nitro, 2-fluoro-4-nitrobenzylalcohol, 2-fluoro-4-nitro-phenyl-methanol, 3-fluoro-4-hydroxymethyl nitrobenzene
PubChem CID 17981733
IUPAC Name (2-fluoro-4-nitrophenyl)methanol
SMILES OCC1=C(F)C=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 103°C to 105°C
Color White to Yellow
Quantity 1 g
Formula Weight 171.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-4-nitrobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only