Learn More
2-Fluoro-4-nitrobenzoic acid, 98%
CAS: 403-24-7 | C7H4FNO4 | 185.11 g/mol
$166.66 - $582.55
Chemical Identifiers
| CAS | 403-24-7 |
|---|---|
| Molecular Formula | C7H4FNO4 |
| Molecular Weight (g/mol) | 185.11 |
| MDL Number | MFCD00275565 |
| InChI Key | MMWFMFZFCKADEL-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-nitrobenzoic acid, 2-fluoro-4-nitrobenzenecarboxylic acid, 2-fluoro-4-nitrobenzoicacid, 2-fluoro-4-nitro-benzoic acid, 4-carboxy-3-fluoronitrobenzene, benzoic acid, 2-fluoro-4-nitro, pubchem1310, acmc-1cts9, 2-fluoro4-nitrobenzoic acid, 2-fluor-4-nitro-benzoesaure |
| PubChem CID | 302679 |
| SMILES | C1=CC(=C(C=C1[N+](=O)[O-])F)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1977003
|
Thermo Scientific Chemicals
L1977003 |
1 g |
Each for $166.66
|
|
|||||
|
AAL1977006
|
Thermo Scientific Chemicals
L1977006 |
5 g |
Each for $582.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 403-24-7 | |
| 185.11 | |
| MMWFMFZFCKADEL-UHFFFAOYSA-N | |
| 302679 |
| C7H4FNO4 | |
| MFCD00275565 | |
| 2-fluoro-4-nitrobenzoic acid, 2-fluoro-4-nitrobenzenecarboxylic acid, 2-fluoro-4-nitrobenzoicacid, 2-fluoro-4-nitro-benzoic acid, 4-carboxy-3-fluoronitrobenzene, benzoic acid, 2-fluoro-4-nitro, pubchem1310, acmc-1cts9, 2-fluoro4-nitrobenzoic acid, 2-fluor-4-nitro-benzoesaure | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)C(=O)O |
Specifications
| 403-24-7 | |
| C7H4FNO4 | |
| 1 g | |
| 2-fluoro-4-nitrobenzoic acid, 2-fluoro-4-nitrobenzenecarboxylic acid, 2-fluoro-4-nitrobenzoicacid, 2-fluoro-4-nitro-benzoic acid, 4-carboxy-3-fluoronitrobenzene, benzoic acid, 2-fluoro-4-nitro, pubchem1310, acmc-1cts9, 2-fluoro4-nitrobenzoic acid, 2-fluor-4-nitro-benzoesaure | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)C(=O)O | |
| 302679 | |
| 98% |
| 172°C to 175°C | |
| MFCD00275565 | |
| 2582091 | |
| MMWFMFZFCKADEL-UHFFFAOYSA-N | |
| 185.11 | |
| 185.11 | |
| 2-Fluoro-4-nitrobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only