missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroanisole 98.0+%, TCI America™

$44.28 - $123.21

Chemical Identifiers

CAS 455-93-6
Molecular Formula C7H6FNO3
Molecular Weight (g/mol) 171.127
MDL Number MFCD00061095
InChI Key XGMVTXUXZUPGGY-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitroanisole, 3-fluoro-4-methoxynitrobenzene, benzene, 2-fluoro-1-methoxy-4-nitro, 2-fluoro-1-methoxy-4-nitro-benzene, pharmabridge p-3000, 2-fluor-1-methoxy-4-nitrobenzol, pubchem2145, 2-fluoro-4-nitroanisol, 4-nitro-2-fluoroanisol, acmc-1apvd
PubChem CID 223104
IUPAC Name 2-fluoro-1-methoxy-4-nitrobenzene
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F06171G
View Documents
TCI America
F06171G
1 g
Each for $44.28
Only null left
 
F06175G
View Documents
TCI America
F06175G
5 g
Each for $123.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

455-93-6
171.127
XGMVTXUXZUPGGY-UHFFFAOYSA-N
223104
COC1=C(C=C(C=C1)[N+](=O)[O-])F
C7H6FNO3
MFCD00061095
2-fluoro-4-nitroanisole, 3-fluoro-4-methoxynitrobenzene, benzene, 2-fluoro-1-methoxy-4-nitro, 2-fluoro-1-methoxy-4-nitro-benzene, pharmabridge p-3000, 2-fluor-1-methoxy-4-nitrobenzol, pubchem2145, 2-fluoro-4-nitroanisol, 4-nitro-2-fluoroanisol, acmc-1apvd
2-fluoro-1-methoxy-4-nitrobenzene

Specifications

455-93-6
C7H6FNO3
1 g
XGMVTXUXZUPGGY-UHFFFAOYSA-N
2-fluoro-1-methoxy-4-nitrobenzene
223104
≥98.0% (GC)
2-Fluoro-4-nitroanisole
105°C
MFCD00061095
2-fluoro-4-nitroanisole, 3-fluoro-4-methoxynitrobenzene, benzene, 2-fluoro-1-methoxy-4-nitro, 2-fluoro-1-methoxy-4-nitro-benzene, pharmabridge p-3000, 2-fluor-1-methoxy-4-nitrobenzol, pubchem2145, 2-fluoro-4-nitroanisol, 4-nitro-2-fluoroanisol, acmc-1apvd
COC1=C(C=C(C=C1)[N+](=O)[O-])F
171.127
171.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'