missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-methylpyridine 98.0+%, TCI America™
Supplier: TCI America F057925G
Specifications
| 2-Fluoro-4-methylpyridine | |
| Yellow | |
| C6H6FN | |
| 25 g | |
| 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j | |
| CC1=CC(=NC=C1)F | |
| 111.119 | |
| 111.12 | |
| Liquid |
| 461-87-0 | |
| 157°C | |
| MFCD00041224 | |
| 1993 | |
| ZBFAXMKJADVOGH-UHFFFAOYSA-N | |
| 2-fluoro-4-methylpyridine | |
| 2737403 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 461-87-0 | |
| 111.119 | |
| ZBFAXMKJADVOGH-UHFFFAOYSA-N | |
| 2737403 | |
| CC1=CC(=NC=C1)F |
| C6H6FN | |
| MFCD00041224 | |
| 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j | |
| 2-fluoro-4-methylpyridine |
Safety and Handling
TSCA : No