missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylpyridine 98.0+%, TCI America™

Catalog No. p-7129766
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05795G 5 g
F057925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05795G Supplier TCI America Supplier No. F05795G
Only null left

Chemical Identifiers

CAS 461-87-0
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD00041224
InChI Key ZBFAXMKJADVOGH-UHFFFAOYSA-N
Synonym 2-fluoro-4-picoline, 2-fluoro-4-methyl-pyridine, pyridine, 2-fluoro-4-methyl, 2-fluoro-4-methyl pyridine, pubchem1203, acmc-1aevi, 4-methyl-2-fluoropyridine, 2-fluoro-4-methylpyridine,, ksc494a2j
PubChem CID 2737403
IUPAC Name 2-fluoro-4-methylpyridine
SMILES CC1=CC(=NC=C1)F

Specifications

Color Yellow
Boiling Point 157°C
Quantity 5 g
UN Number 1993
Formula Weight 111.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-4-methylpyridine