missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-methylbenzonitrile 98.0+%, TCI America™
$63.38 - $192.00
Chemical Identifiers
| CAS | 85070-67-3 |
|---|---|
| Molecular Formula | C8H6FN |
| Molecular Weight (g/mol) | 135.14 |
| MDL Number | MFCD03094311 |
| InChI Key | WCGNLBCJPBKXCN-UHFFFAOYSA-N |
| Synonym | 4-cyano-3-fluorotoluene, 2-fluoro-4-methylbenzenecarbonitrile, 2-fluoro-4-methyl-benzonitrile, benzonitrile, 2-fluoro-4-methyl, pubchem1555, 2-fluoro-p-tolunitrile, acmc-209q3w, ksc494e6d, 4-methyl-2-fluorobenzonitrile, pharmabridge p-1079 |
| PubChem CID | 2778443 |
| IUPAC Name | 2-fluoro-4-methylbenzonitrile |
| SMILES | CC1=CC=C(C#N)C(F)=C1 |
Chemical Identifiers
| 85070-67-3 | |
| 135.14 | |
| WCGNLBCJPBKXCN-UHFFFAOYSA-N | |
| 2778443 | |
| CC1=CC=C(C#N)C(F)=C1 |
| C8H6FN | |
| MFCD03094311 | |
| 4-cyano-3-fluorotoluene, 2-fluoro-4-methylbenzenecarbonitrile, 2-fluoro-4-methyl-benzonitrile, benzonitrile, 2-fluoro-4-methyl, pubchem1555, 2-fluoro-p-tolunitrile, acmc-209q3w, ksc494e6d, 4-methyl-2-fluorobenzonitrile, pharmabridge p-1079 | |
| 2-fluoro-4-methylbenzonitrile |
Specifications
| 85070-67-3 | |
| White-Yellow | |
| MFCD03094311 | |
| 3439 | |
| WCGNLBCJPBKXCN-UHFFFAOYSA-N | |
| 2-fluoro-4-methylbenzonitrile | |
| 2778443 | |
| ≥98.0% (GC) | |
| 2-Fluoro-4-methylbenzonitrile |
| 53°C | |
| C8H6FN | |
| 1 g | |
| 4-cyano-3-fluorotoluene, 2-fluoro-4-methylbenzenecarbonitrile, 2-fluoro-4-methyl-benzonitrile, benzonitrile, 2-fluoro-4-methyl, pubchem1555, 2-fluoro-p-tolunitrile, acmc-209q3w, ksc494e6d, 4-methyl-2-fluorobenzonitrile, pharmabridge p-1079 | |
| CC1=CC=C(C#N)C(F)=C1 | |
| 135.14 | |
| 135.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No