missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-methylbenzoic Acid 98.0+%, TCI America™
$80.16 - $238.60
Chemical Identifiers
| CAS | 7697-23-6 |
|---|---|
| Molecular Formula | C8H7FO2 |
| Molecular Weight (g/mol) | 154.14 |
| MDL Number | MFCD03092906 |
| InChI Key | ALFWHEYHCZRVLO-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-methyl-benzoic acid, 2-fluoro-p-toluic acid, 2-fluoro-4-methylbenzoicacid, benzoic acid, 2-fluoro-4-methyl, pubchem1395, acmc-1bbau, 4-carboxy-3-fluorotoluene, ksc377a4r, rarechem al bo 2237, 2-fluoro-4-methyl benzoic acid |
| PubChem CID | 2736145 |
| IUPAC Name | 2-fluoro-4-methylbenzoic acid |
| SMILES | CC1=CC=C(C(O)=O)C(F)=C1 |
Chemical Identifiers
| 7697-23-6 | |
| 154.14 | |
| ALFWHEYHCZRVLO-UHFFFAOYSA-N | |
| 2736145 | |
| CC1=CC=C(C(O)=O)C(F)=C1 |
| C8H7FO2 | |
| MFCD03092906 | |
| 2-fluoro-4-methyl-benzoic acid, 2-fluoro-p-toluic acid, 2-fluoro-4-methylbenzoicacid, benzoic acid, 2-fluoro-4-methyl, pubchem1395, acmc-1bbau, 4-carboxy-3-fluorotoluene, ksc377a4r, rarechem al bo 2237, 2-fluoro-4-methyl benzoic acid | |
| 2-fluoro-4-methylbenzoic acid |
Specifications
| 7697-23-6 | |
| C8H7FO2 | |
| 1 g | |
| ALFWHEYHCZRVLO-UHFFFAOYSA-N | |
| 2-fluoro-4-methylbenzoic acid | |
| 2736145 | |
| ≥98.0% (GC,T) | |
| 2-Fluoro-4-methylbenzoic Acid |
| 186°C | |
| MFCD03092906 | |
| 2-fluoro-4-methyl-benzoic acid, 2-fluoro-p-toluic acid, 2-fluoro-4-methylbenzoicacid, benzoic acid, 2-fluoro-4-methyl, pubchem1395, acmc-1bbau, 4-carboxy-3-fluorotoluene, ksc377a4r, rarechem al bo 2237, 2-fluoro-4-methyl benzoic acid | |
| CC1=CC=C(C(O)=O)C(F)=C1 | |
| 154.14 | |
| 154.14 | |
| Crystalline Powder |
Safety and Handling
TSCA : No