missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. F08131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08131G 1 g
F08135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08131G Supplier TCI America Supplier No. F08131G

Chemical Identifiers

CAS 7697-23-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD03092906
InChI Key ALFWHEYHCZRVLO-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyl-benzoic acid, 2-fluoro-p-toluic acid, 2-fluoro-4-methylbenzoicacid, benzoic acid, 2-fluoro-4-methyl, pubchem1395, acmc-1bbau, 4-carboxy-3-fluorotoluene, ksc377a4r, rarechem al bo 2237, 2-fluoro-4-methyl benzoic acid
PubChem CID 2736145
IUPAC Name 2-fluoro-4-methylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(F)=C1

Specifications

Melting Point 186°C
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-methylbenzoic Acid