missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methylbenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. AAH5912603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5912603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5912603 Supplier Thermo Scientific Chemicals Supplier No. H5912603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1165936-03-7
Molecular Formula C13H18BFO2
Molecular Weight (g/mol) 236.093
MDL Number MFCD18779806
InChI Key MKWDECBHRFNFGY-UHFFFAOYSA-N
Synonym 2-2-fluoro-4-methylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-4-methylphenylboronic acid pinacol ester, 2-fluoro-4-methylbenzeneboronic acid pinacol ester, 2-fluoro-4-methylphenyl boronic acid pinacol ester
PubChem CID 69672435
IUPAC Name 2-(2-fluoro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C)F

Specifications

Melting Point 58°C to 60°C
Quantity 1 g
Formula Weight 236.09
Percent Purity 96%
Chemical Name or Material 2-Fluoro-4-methylbenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures