missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%, Thermo Scientific™
Specifications
Specifications
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Quantity | 5g |
| Formula Weight | 197.96 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Fluoro-4-methoxycarbonylphenylboronic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only