missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%, Thermo Scientific™

Catalog No. p-7077585
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455870050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC455870050 Supplier Thermo Scientific Chemicals Supplier No. 455870050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 603122-84-5
Molecular Formula C8H8BFO4
Molecular Weight (g/mol) 197.96
InChI Key BKWRLCIYMAYFPA-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
PubChem CID 24824345
IUPAC Name (2-fluoro-4-methoxycarbonylphenyl)boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)OC)F)(O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5g
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methoxycarbonylphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only