missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F10711G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F10711G Supplier TCI America Supplier No. F10711G

Specifications

Chemical Name or Material 2-Fluoro-4-(methoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 603122-84-5
Melting Point 140°C
Color White
Molecular Formula C8H8BFO4
Quantity 1 g
MDL Number MFCD08436034
Synonym 2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
InChI Key BKWRLCIYMAYFPA-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(F)=C(C=C1)B(O)O
IUPAC Name [2-fluoro-4-(methoxycarbonyl)phenyl]boronic acid
Molecular Weight (g/mol) 197.96
PubChem CID 24824345
Formula Weight 197.96
Physical Form Crystalline Powder
Show More Show Less