missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxybenzyl alcohol, 97%

Catalog No. AAH3211809
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3211809 10 g
AAH3211803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3211809 Supplier Thermo Scientific Chemicals Supplier No. H3211809
Only null left

CAS: 405-09-4 | C8H9FO2 | 156.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 405-09-4
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.16
MDL Number MFCD04116336
InChI Key AZYGOIQKPGPBTM-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxyphenyl methanol, 2-fluoro-4-methoxybenzyl alcohol, 2-fluoro-p-anisyl alcohol, 3-fluoro-4-hydroxymethyl anisole, benzenemethanol,2-fluoro-4-methoxy, 2-fluoro-4-methoxyphenyl methan-1-ol, benzyl alcohol, 4-methoxy-6-fluoro, pubchem7733, 2-fluoro-4-methoxybenzylalcohol, rarechem al bd 0456
PubChem CID 593638
IUPAC Name (2-fluoro-4-methoxyphenyl)methanol
SMILES COC1=CC(F)=C(CO)C=C1

Specifications

Quantity 10 g
Formula Weight 156.15
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxybenzyl alcohol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only