missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Fluoro-4'-methoxyacetophenone 98.0+%, TCI America™
Supplier: TCI America F06625G
Specifications
| 2′-Fluoro-4′-methoxyacetophenone | |
| 52°C | |
| 85°C | |
| MFCD00042290 | |
| 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344 | |
| COC1=CC=C(C(C)=O)C(F)=C1 | |
| 168.17 | |
| 168.17 | |
| Crystalline Powder |
| 74457-86-6 | |
| White-Yellow | |
| C9H9FO2 | |
| 5 g | |
| PIRRWUMTIBFCCW-UHFFFAOYSA-N | |
| 1-(2-fluoro-4-methoxyphenyl)ethan-1-one | |
| 592821 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 74457-86-6 | |
| 168.17 | |
| PIRRWUMTIBFCCW-UHFFFAOYSA-N | |
| 592821 | |
| COC1=CC=C(C(C)=O)C(F)=C1 |
| C9H9FO2 | |
| MFCD00042290 | |
| 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344 | |
| 1-(2-fluoro-4-methoxyphenyl)ethan-1-one |
Safety and Handling
TSCA : No