missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-methoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7129819
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06621G 1 g
F06625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06621G Supplier TCI America Supplier No. F06621G
Only null left

Chemical Identifiers

CAS 74457-86-6
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD00042290
InChI Key PIRRWUMTIBFCCW-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-methoxyacetophenone, 2-fluoro-4-methoxyacetophenone, 1-2-fluoro-4-methoxyphenyl ethanone, 1-2-fluoro-4-methoxyphenyl ethan-1-one, 4-acetyl-3-fluoroanisole, 1-acetyl-2-fluoro-4-methoxybenzene, 2'-fluor-4'-methoxyacetophenone, 2-fluoro-4-methoxy-acetophenone, ethanone, 1-2-fluoro-4-methoxyphenyl, zlchem 344
PubChem CID 592821
IUPAC Name 1-(2-fluoro-4-methoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C(C)=O)C(F)=C1

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 85°C
Quantity 1 g
Formula Weight 168.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Fluoro-4′-methoxyacetophenone