missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-iodotoluene 95.0+%, TCI America™
Supplier: TCI America F072025G
Specifications
| 2-Fluoro-4-iodotoluene | |
| Yellow | |
| C7H6FI | |
| 25 g | |
| QZLWTFTXGKKCHZ-UHFFFAOYSA-N | |
| 2-fluoro-4-iodo-1-methylbenzene | |
| 123492 | |
| ≥95.0% (GC) |
| 39998-81-7 | |
| 208°C | |
| MFCD00001045 | |
| 2-fluoro-4-iodotoluene, 2-fluoro-4-iodo-1-methyl-benzene, benzene, 2-fluoro-4-iodo-1-methyl, 2-fluor-4-iod-1-methylbenzol, pubchem1618, 2-fluoro-4-iodo-toluene, acmc-209j9t, 3-fluoro-4-methyl-iodobenzene, buttpark 50\01-77, # | |
| CC1=C(C=C(C=C1)I)F | |
| 236.028 | |
| 236.03 | |
| Liquid |
Chemical Identifiers
| 39998-81-7 | |
| 236.028 | |
| QZLWTFTXGKKCHZ-UHFFFAOYSA-N | |
| 123492 | |
| CC1=C(C=C(C=C1)I)F |
| C7H6FI | |
| MFCD00001045 | |
| 2-fluoro-4-iodotoluene, 2-fluoro-4-iodo-1-methyl-benzene, benzene, 2-fluoro-4-iodo-1-methyl, 2-fluor-4-iod-1-methylbenzol, pubchem1618, 2-fluoro-4-iodo-toluene, acmc-209j9t, 3-fluoro-4-methyl-iodobenzene, buttpark 50\01-77, # | |
| 2-fluoro-4-iodo-1-methylbenzene |
Safety and Handling
TSCA : No