missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-iodo-5-methylpyridine 98.0+%, TCI America™

Catalog No. F10351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10351G 1 g
F10355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10351G Supplier TCI America Supplier No. F10351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 153034-94-7
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD03095297
InChI Key BVKQLNXPPQEELX-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-5-picoline, 2-fluoro-4-iodo-5-methyl-pyridine, 6-fluoro-4-iodo-b-picoline, 6-fluoro-4-iodo-3-picoline, 2-fluoro-4-iodo-5-methyl pyridine, pyridine,2-fluoro-4-iodo-5-methyl, pyridine, 2-fluoro-4-iodo-5-methyl, pubchem1305, acmc-20el7f, abbypharma ap-30-1289
PubChem CID 10060052
IUPAC Name 2-fluoro-4-iodo-5-methylpyridine
SMILES CC1=CN=C(C=C1I)F

Specifications

Melting Point 44°C
Quantity 1 g
Formula Weight 237.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-iodo-5-methylpyridine