missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzonitrile 98.0+%, TCI America™

Catalog No. p-7129701
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04625G 5 g
F046225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04625G Supplier TCI America Supplier No. F04625G

Chemical Identifiers

CAS 82380-18-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00192177
InChI Key REIVHYDACHXPNH-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorophenol, 3-fluoro-4-cyanophenol, 2-fluoro-4-benzonitrile, benzonitrile, 2-fluoro-4-hydroxy, 4-hydroxy-2-fluorobenzonitrile, 2-fluoro-4-hydroxy-benzonitrile, 2-fluoro-4-hydroxybenzenecarbonitrile, pubchem2610, pubchem3495
PubChem CID 2734675
IUPAC Name 2-fluoro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1O)F)C#N

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 137.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-hydroxybenzonitrile
EINECSNumber (3)-4547
TSCA No