missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzonitrile, 99%

Catalog No. AAB2153503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2153503 1 g
AAB2153506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2153503 Supplier Thermo Scientific Chemicals Supplier No. B2153503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82380-18-5 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 82380-18-5
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00192177
InChI Key REIVHYDACHXPNH-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorophenol, 3-fluoro-4-cyanophenol, 2-fluoro-4-benzonitrile, benzonitrile, 2-fluoro-4-hydroxy, 4-hydroxy-2-fluorobenzonitrile, 2-fluoro-4-hydroxy-benzonitrile, 2-fluoro-4-hydroxybenzenecarbonitrile, pubchem2610, pubchem3495
PubChem CID 2734675
IUPAC Name 2-fluoro-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1O)F)C#N

Specifications

Melting Point 123°C to 126°C
Density 1.45
Quantity 1 g
UN Number UN3077
Beilstein 4741066
Formula Weight 137.11
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-hydroxybenzonitrile
Hazard Category H302-H318
Hazard Statement GHS H Statement
H301-H318
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 422-810-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only