missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™
$82.02 - $329.94
Chemical Identifiers
| CAS | 65145-13-3 |
|---|---|
| Molecular Formula | C7H5FO3 |
| Molecular Weight (g/mol) | 156.11 |
| MDL Number | MFCD01310985 |
| InChI Key | NXWTWYULZRDBSA-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid |
| PubChem CID | 2783398 |
| IUPAC Name | 2-fluoro-4-hydroxybenzoic acid |
| SMILES | OC(=O)C1=C(F)C=C(O)C=C1 |
Chemical Identifiers
| 65145-13-3 | |
| 156.11 | |
| NXWTWYULZRDBSA-UHFFFAOYSA-N | |
| 2783398 | |
| OC(=O)C1=C(F)C=C(O)C=C1 |
| C7H5FO3 | |
| MFCD01310985 | |
| 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid | |
| 2-fluoro-4-hydroxybenzoic acid |
Specifications
| 65145-13-3 | |
| White-Yellow | |
| MFCD01310985 | |
| 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid | |
| OC(=O)C1=C(F)C=C(O)C=C1 | |
| 156.11 | |
| 156.11 | |
| Crystalline Powder |
| 202°C | |
| C7H5FO3 | |
| 5 g | |
| NXWTWYULZRDBSA-UHFFFAOYSA-N | |
| 2-fluoro-4-hydroxybenzoic acid | |
| 2783398 | |
| ≥98.0% (GC,T) | |
| 2-Fluoro-4-hydroxybenzoic Acid |
Safety and Handling
TSCA : No