missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzaldehyde, 97%

Catalog No. AAH26218MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26218MC 100 mg
AAH26218ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH26218MC Supplier Thermo Scientific Chemicals Supplier No. H26218MC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 348-27-6 | C7H5FO2 | 140.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 348-27-6
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD06797918
InChI Key ONRPXRPUBXXCCM-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzaldehyde, benzaldehyde, 2-fluoro-4-hydroxy, pubchem1442, acmc-1act5, ksc497m4n, #
PubChem CID 587246
IUPAC Name 2-fluoro-4-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1O)F)C=O

Specifications

Melting Point 168°C to 170°C
Quantity 100 mg
Sensitivity Air Sensitive
Formula Weight 140.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-hydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only