CAS RN 348-27-6
IUPAC Name:
2-fluoro-4-hydroxybenzaldehyde
Synonyms:
2-fluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol):
140.11
Molecular Formula:
C7H5FO2
InChi Key:
ONRPXRPUBXXCCM-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=C(C=O)C=C1
1
–
1
of
1
results
1
Promotions Available
2-Fluoro-4-hydroxybenzaldehyde, 97%
2-Fluoro-4-hydroxybenzaldehyde, 97%
CAS: 348-27-6 Molecular Formula: C7H5FO2 Molecular Weight (g/mol): 140.113 MDL Number: MFCD06797918 InChI Key: ONRPXRPUBXXCCM-UHFFFAOYSA-N Synonym: 2-fluoro-4-hydroxy-benzaldehyde,benzaldehyde, 2-fluoro-4-hydroxy,pubchem1442,acmc-1act5,ksc497m4n,# PubChem CID: 587246 IUPAC Name: 2-fluoro-4-hydroxybenzaldehyde SMILES: C1=CC(=C(C=C1O)F)C=O