missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-4'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. F04181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04181G 1 g
F04185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04181G Supplier TCI America Supplier No. F04181G

Chemical Identifiers

CAS 98619-07-9
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00142713
InChI Key ZCZIRBNZMFUCOH-UHFFFAOYSA-N
Synonym 2'-fluoro-4'-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl ethanone, 4-hydroxy-2-fluoroacetophenone, 2-fluoro-4-hydroxyacetophenone, 1-2-fluoro-4-hydroxyphenyl-1-ethanone, 1-2-fluoro-4-hydroxyphenyl ethan-1-one, 1-2-fluoro-4-hydroxy-phenyl ethanone, ethanone, 1-2-fluoro-4-hydroxyphenyl, 4'-hydroxy-2'-fluoroacetophenone
PubChem CID 2724912
IUPAC Name 1-(2-fluoro-4-hydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C=C1F

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Fluoro-4′-hydroxyacetophenone