missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4,5-dimethoxybenzonitrile 98.0+%, TCI America™
$141.67 - $421.27
Chemical Identifiers
| CAS | 119396-88-2 |
|---|---|
| Molecular Formula | C9H8FNO2 |
| Molecular Weight (g/mol) | 181.166 |
| MDL Number | MFCD00143236 |
| InChI Key | QZKGCLSVCWWARO-UHFFFAOYSA-N |
| Synonym | 4-Cyano-5-fluoroveratrole |
| PubChem CID | 2774258 |
| IUPAC Name | 2-fluoro-4,5-dimethoxybenzonitrile |
| SMILES | COC1=C(C=C(C(=C1)C#N)F)OC |
Chemical Identifiers
| 119396-88-2 | |
| 181.166 | |
| QZKGCLSVCWWARO-UHFFFAOYSA-N | |
| 2774258 | |
| COC1=C(C=C(C(=C1)C#N)F)OC |
| C9H8FNO2 | |
| MFCD00143236 | |
| 4-Cyano-5-fluoroveratrole | |
| 2-fluoro-4,5-dimethoxybenzonitrile |
Specifications
| 119396-88-2 | |
| C9H8FNO2 | |
| 1 g | |
| 4-Cyano-5-fluoroveratrole | |
| COC1=C(C=C(C(=C1)C#N)F)OC | |
| 181.166 | |
| 181.17 | |
| Crystalline Powder |
| 143°C | |
| MFCD00143236 | |
| 3439 | |
| QZKGCLSVCWWARO-UHFFFAOYSA-N | |
| 2-fluoro-4,5-dimethoxybenzonitrile | |
| 2774258 | |
| ≥98.0% (GC) | |
| 2-Fluoro-4,5-dimethoxybenzonitrile |
Safety and Handling
TSCA : No