missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4,5-dimethoxybenzonitrile 98.0+%, TCI America™

Catalog No. F06415G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06415G 5 g
F06411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06415G Supplier TCI America Supplier No. F06415G
Only null left

Chemical Identifiers

CAS 119396-88-2
Molecular Formula C9H8FNO2
Molecular Weight (g/mol) 181.166
MDL Number MFCD00143236
InChI Key QZKGCLSVCWWARO-UHFFFAOYSA-N
Synonym 4-Cyano-5-fluoroveratrole
PubChem CID 2774258
IUPAC Name 2-fluoro-4,5-dimethoxybenzonitrile
SMILES COC1=C(C=C(C(=C1)C#N)F)OC

Specifications

Melting Point 143°C
Quantity 5 g
UN Number 3439
Formula Weight 181.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4,5-dimethoxybenzonitrile