missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4,5-dimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. f04411g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04411G 1 g
F04415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04411G Supplier TCI America Supplier No. F04411G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71924-62-4
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD00061108
InChI Key IBBYQNVXKFMSSI-UHFFFAOYSA-N
PubChem CID 603595
IUPAC Name 2-fluoro-4,5-dimethoxybenzaldehyde
SMILES COC1=CC(F)=C(C=O)C=C1OC

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Synonym 6-fluoroveratraldehyde, 4,5-dimethoxy-2-fluorobenzaldehyde, benzaldehyde,2-fluoro-4,5-dimethoxy, benzaldehyde, 2-fluoro-4,5-dimethoxy, pubchem2651, acmc-1bkgo, 4.5-dimethoxy-2-fluorobenzaldehyde, 2-fluoro-4,5-dimethoxy-benzaldehyde, 4,5-dimethoxy-2-fluoro benzaldehyde
Formula Weight 184.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4,5-dimethoxybenzaldehyde