missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-(trifluoromethyl)benzoic acid, 98%

Catalog No. AAB2028003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2028003 1 g
AAB2028006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2028003 Supplier Thermo Scientific Chemicals Supplier No. B2028003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 115029-22-6 | C8H3F4O2 | 207.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115029-22-6
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040980
InChI Key XVEAMDNSCPPPCP-UHFFFAOYSA-M
Synonym 2-fluoro-3-trifluoromethyl benzoic acid, 2-fluoro-3-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-m-toluic acid, 2-fluoro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-fluoro-3-trifluoromethyl, 2-fluoro-3-trifluoromethylbenzioc acid, pubchem2741, acmc-1boz3, ksc489q1l, rarechem al bo 0625
PubChem CID 518002
IUPAC Name 2-fluoro-3-(trifluoromethyl)benzoic acid
SMILES [O-]C(=O)C1=CC=CC(=C1F)C(F)(F)F

Specifications

Melting Point 126°C to 128°C
Quantity 1 g
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-3-(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only